Vinyltrimethoxysilane 98%

Supplier: Thermo Scientific Chemicals
Warning

B21176-22 B21176-36
CAAAB21176-22EA 78.68 CAD
CAAAB21176-22 CAAAB21176-36
Vinyltrimethoxysilane 98%
Vinyltrimethoxysilane

Purity: 98% CAS No.: 2768-02-7 Molecular Formula: C5H12O3Si Formula Weight: 148.24 Flash Point: 23°(73°F) Refractive Index: 1.3920 Odor: Fruit-like Melting Point: -40° Boiling Point: 122-124° Beilstein Reference: 1099136 UN No.: 1993 MDL No.: MFCD00008605

Formula: C₅H₁₂O₃Si
Boiling Pt: 122…124 °C
Melting Pt: < –40 °C
Density: 0.970
Flash Pt: 23 °C(73 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008605
CAS Number: 2768-02-7
EINECS: 220-449-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR