Molecular sieve 3A (0.3 nm, 3 Å), powder
Supplier: Thermo Scientific Chemicals
Synonyms:
Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate
B21165-0B
B21165-0I
B21165-30
CAAAB21165-0BEA
150.42
CAD
CAAAB21165-0B
CAAAB21165-0I
CAAAB21165-30
Molecular sieve 3A (0.3 nm, 3 Å), powder
Molecular sieve 3A (0.3 nm, 3 Å)
CAS No.: 308080-99-1
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Pale brown
Odor: Odorless
MDL No.: MFCD00147627
Formula:
KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00131613 CAS Number: 308080-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...