IPTG (isopropyl-β-D-thiogalactopyranoside) 99% dioxane-free
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside
B21149-03
B21149-06
CAAAB21149-03EA
139.99
CAD
CAAAB21149-03
CAAAB21149-06
IPTG (isopropyl-β-D-thiogalactopyranoside) 99% dioxane-free
IPTG (Isopropyl-β-D-thiogalactopyranoside)
Beta-galactosidase inducer
Purity: 99%
CAS No.: 367-93-1
Molecular Formula: C9H18O5S
Formula Weight: 238.30
Storage Temperature: -30°C to -10°C
Melting Point: 109-115°
Beilstein Reference: 4631
Optical Rotation: -32° (c=1 in water)
MDL No.: MFCD00063273
Formula:
C₉H₁₈O₅S MW: 238.31 g/mol Boiling Pt: 402 Melting Pt: 109…115 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063273 CAS Number: 367-93-1 EINECS: 206-703-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...