Methyl yellow ≥98% indicator
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21145-14
B21145-22
B21145-36
CAAAB21145-14EA
48.82
CAD
CAAAB21145-14
CAAAB21145-22
CAAAB21145-36
Methyl yellow ≥98% indicator
Methyl Yellow
CAS No.: 60-11-7
Molecular Formula: C14H15N3
Formula Weight: 225.30
Odor: Odorless
Melting Point: 110-112° dec.
Solubility: Insoluble in water. Soluble in alcohol.
Beilstein Reference: 746016
Merck Reference: 14,3229
UN No.: 2811
MDL No.: MFCD00008308
Formula:
C₁₄H₁₅N₃ Boiling Pt: 371 °C (1013 hPa) Melting Pt: 110…112 °C (dec.) Density: 1.2233 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008308 CAS Number: 60-11-7 EINECS: 200-455-7 UN: 3143 ADR: 6.1,III Merck Index: 14,3229 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...