5,6-Dibromovanillin 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,3-Dibromo-4-hydroxy-5-methoxybenzaldehyde
B21140-03
B21140-06
B21140-14
CAAAB21140-03EA
46.93
CAD
CAAAB21140-03
CAAAB21140-06
CAAAB21140-14
5,6-Dibromovanillin 98%
5,6-Dibromovanillin
Purity: 98%
CAS No.: 2973-75-3
Molecular Formula: C8H6Br2O3
Formula Weight: 309.94
Melting Point: 213-217°
Beilstein Reference: 1968576
Formula:
C₈H₆Br₂O₃ Melting Pt: 213…217 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016978 CAS Number: 2973-75-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...