(3,4,5-Trimethoxyphenyl)acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

B21136-06 B21136-14 B21136-22
CAAAB21136-06EA 79.63 CAD
CAAAB21136-06 CAAAB21136-14 CAAAB21136-22
(3,4,5-Trimethoxyphenyl)acetic acid 99%
(3,4,5-Trimethoxyphenyl)acetic acid

Purity: 99% CAS No.: 951-82-6 Molecular Formula: C11H14O5 Formula Weight: 226.23 Melting Point: 118-122° Beilstein Reference: 2697844

Formula: (C₆H₂(OCH₃)₃)CH₂COOH
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00004336
CAS Number: 951-82-6
EINECS: 213-456-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR