Molecular sieve 13X (1.0 nm, 10 Å), beads 1.6-2.5 mm (0.063-0.098 in)

Supplier: Thermo Scientific Chemicals

B21109-0B B21109-0I B21109-30
CAAAB21109-0BEA 195.92 CAD
CAAAB21109-0B CAAAB21109-0I CAAAB21109-30
Molecular sieve 13X (1.0 nm, 10 Å), beads 1.6-2.5 mm (0.063-0.098 in)
Molecular sieve 13X (1.0 nm, 10 Å)

CAS No.: 63231-69-6 MDL No.: MFCD00131613

Storage Temperature: Ambient
MDL Number: MFCD00131613
CAS Number: 63231-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR