2-Methoxyphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21071-14
B21071-03
B21071-06
CAAAAB21071-14EA
480.32
CAD
CAAAAB21071-14
CAAAB21071-03
CAAAB21071-06
2-Methoxyphenylboronic acid 97%
2-Methoxyphenylboronic acid
Purity: 97%
CAS No.: 5720-06-9
Molecular Formula: C7H9BO3
Formula Weight: 151.96
Melting Point: 102-106°
Beilstein Reference: 3030981
Formula:
C₇H₉BO₃ Melting Pt: 102…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236047 CAS Number: 5720-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...