Isobutyltrimethoxysilane 97%

Supplier: Thermo Scientific Chemicals
Warning

B21048-14 B21048-22 B21048-36
CAAAB21048-14EA 37.76 CAD
CAAAB21048-14 CAAAB21048-22 CAAAB21048-36
Isobutyltrimethoxysilane 97%
Isobutyltrimethoxysilane

Purity: 97% CAS No.: 18395-30-7 Molecular Formula: (CH3)2CHCH2Si(OCH3)3 Formula Weight: 178.31 Flash Point: 43°(109°F) Refractive Index: 1.3960 Boiling Point: 154-157° Beilstein Reference: 2233976 UN No.: 1993 MDL No.: MFCD00051589

Formula: (H₃C)₂HCSi(OCH₃)₃
Boiling Pt: 154…157 °C
Density: 0.930
Flash Pt: 43 °C (109 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051589
CAS Number: 18395-30-7
EINECS: 242-272-5
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR