5-Chloro-2-methylindole 98%

Supplier: Thermo Scientific Chemicals

B21036-03 B21036-06
CAAAB21036-03EA 61.46 CAD
CAAAB21036-03 CAAAB21036-06 CAAAB21036-14
5-Chloro-2-methylindole 98%
5-Chloro-2-methylindole

Purity: 98% CAS No.: 1075-35-0 Molecular Formula: C9H8ClN Formula Weight: 165.62 Melting Point: 110-114° MDL No.: MFCD00005619

Formula: C₉H₈ClN
Boiling Pt: 130 - 140 °C (0.1 -rr)
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00005619
CAS Number: 1075-35-0
EINECS: 214-052-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR