X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 98+%

Supplier: Thermo Scientific Chemicals
Warning

B21034-MD B21034-MF
CAAAAB21034-MDEA 369.72 CAD
CAAAAB21034-MD CAAAB21034-MA CAAAB21034-MF
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 98+%
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)

Substrate for beta-galactosidase


Purity: 98+% CAS No.: 7240-90-6 Molecular Formula: C14H15BrClNO6 Formula Weight: 408.64 Storage Temperature: 2°C to 8°C Melting Point: ca 236° dec. Beilstein Reference: 1402009 Optical Rotation: -62° (c=1 in 50% DMF) MDL No.: MFCD00005666

Formula: C₁₄H₁₅BrClNO₆
Melting Pt: ca 236 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005666
CAS Number: 7240-90-6
EINECS: 230-640-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR