Propyltrimethoxysilane 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21033-22
B21033-36
CAAAB21033-22EA
178.54
CAD
CAAAB21033-22
CAAAB21033-36
Propyltrimethoxysilane 98+%
Propyltrimethoxysilane
Purity: 98+%
CAS No.: 1067-25-0
Molecular Formula: CH3(CH2)2Si(OCH3)3
Formula Weight: 164.28
Flash Point: 34°(93°F)
Refractive Index: 1.3900
Boiling Point: 142-143°
Beilstein Reference: 2070458
UN No.: 1993
MDL No.: MFCD00043026
Formula:
C₆H₁₆O₃Si Boiling Pt: 142…143 °C Density: 0.935 Flash Pt: 34 °C (93 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00043026 CAS Number: 1067-25-0 EINECS: 213-926-7 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...