D-(-)-Histidine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (R)-(-)-Histidine, D-3-Imidazol-4-ylalanine, (2R)-(-)-Histidine, H-D-His-OH

B21027-22 B21027-06 B21027-14
CAAAAB21027-22EA 2129.84 CAD
CAAAAB21027-22 CAAAB21027-06 CAAAB21027-14
D-(-)-Histidine 99%
D(-)-Histidine

Purity: 99% CAS No.: 351-50-8 Molecular Formula: C6H9N3O2 Formula Weight: 155.16 Melting Point: ca 285° dec. Beilstein Reference: 84089

Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Melting Pt: 287 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00065963
CAS Number: 351-50-8
EINECS: 206-513-8
Merck Index: 12,04758

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR