Propyltriethoxysilane 97%
Supplier: Thermo Scientific Chemicals
B20997-22
B20997-36
CAAAB20997-22EA
199.08
CAD
CAAAB20997-22
CAAAB20997-36
Propyltriethoxysilane 97%
Propyltriethoxysilane
Purity: 97%
CAS No.: 2550-02-9
Molecular Formula: CH3(CH2)2Si(OCH2CH3)3
Formula Weight: 206.36
Flash Point: 57°(134°F)
Refractive Index: 1.3965
Boiling Point: 179-180°
Beilstein Reference: 1748158
UN No.: 1993
MDL No.: MFCD00026756
Formula:
C₉H₂₂O₃Si |
MDL Number:
MFCD00026756 CAS Number: 2550-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...