1,4-Diacetoxybenzene 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Hydroquinone diacetate, 1,4-Phenylene diacetate, p-Phenylene di(acetate)

B20987-14 B20987-22 B20987-30
CAAAAB20987-14EA 48.51 CAD
CAAAAB20987-14 CAAAAB20987-22 CAAAB20987-30
1,4-Diacetoxybenzene 98%
1,4-Diacetoxybenzene

Purity: 98% CAS No.: 1205-91-0 Molecular Formula: C10H10O4 Formula Weight: 194.19 Melting Point: 119-122° Beilstein Reference: 1912233

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 119…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00011643
CAS Number: 1205-91-0
EINECS: 214-887-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR