2-(Trimethylsilyl)ethanol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20970-06
B20970-14
B20970-22
CAAAB20970-06EA
138.72
CAD
CAAAB20970-06
CAAAB20970-14
CAAAB20970-22
2-(Trimethylsilyl)ethanol 98+%
2-(Trimethylsilyl)ethanol
Purity: 98+%
CAS No.: 2916-68-9
Molecular Formula: (CH3)3SiCH2CH2OH
Formula Weight: 118.25
Flash Point: 50°(122°F)
Refractive Index: 1.4250
Boiling Point: 71-72°/10mm
Beilstein Reference: 1732034
UN No.: 1987
MDL No.: MFCD00002825
Formula:
HOH₂CCH₂Si(CH₃)₃ MW: 118.25 g/mol Boiling Pt: 71…72 °C/10 mm Melting Pt: 0 Density: 0.825 Flash Pt: 50 °C (122 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002825 CAS Number: 2916-68-9 EINECS: 220-844-5 UN: 1987 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...