4-Hydroxy-3-nitropyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Nitro-4-pyridon, 3-Nitro-4-pyridone, 3-Nitropyridin-4-ol, 3-Nitro-4-pyridinol

B20963-06 B20963-14 B20963-22
CAAAB20963-06EA 71.26 CAD
CAAAB20963-06 CAAAB20963-14 CAAAB20963-22
4-Hydroxy-3-nitropyridine 95%
4-Hydroxy-3-nitropyridine

Purity: 95% CAS No.: 5435-54-1 Molecular Formula: C5H4N2O3 Formula Weight: 140.10 Melting Point: ca 280° dec. Beilstein Reference: 1072928

Formula: C₅H₄N₂O₃
MW: 140.1
Melting Pt: ∼ 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00128874
CAS Number: 5435-54-1
EINECS: 611-127-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR