1-Iodoperfluorohexane 98+% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Perfluorohexyl iodide, Perfluoro-n-hexyl iodide, 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-6-iodohexane, Perfluoro-1-iodohexane
Stabilization:
Stabilised with copper
B20958-30
B20958-09
B20958-18
CAAAAB20958-30EA
976.44
CAD
CAAAAB20958-30
CAAAB20958-09
CAAAB20958-18
1-Iodoperfluorohexane 98+% stabilized
1-Iodoperfluorohexane
Purity: 98+%
CAS No.: 355-43-1
Molecular Formula: CF3(CF2)5I
Formula Weight: 445.95
Refractive Index: 1.3290
Melting Point: -45°
Boiling Point: 117°
Beilstein Reference: 1802118
MDL No.: MFCD00001063
Formula:
C₆F₁₃I MW: 445.95 g/mol Boiling Pt: 117 °C Melting Pt: –45 °C Density: 2.063 Storage Temperature: Ambient |
MDL Number:
MFCD00001063 CAS Number: 355-43-1 EINECS: 206-586-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...