m-Anisohydrazide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Methoxybenzohydrazide, 3-Methoxybenzene-1-carbohydrazide, m-Anisic hydrazide, 3-Anisohydrazide, 3-Methoxybenzhydrazide, 3-Methoxybenzoic acid hydrazide, 3-Methoxybenzoic hydrazide
B20956-06
B20956-14
CAAAB20956-06EA
43.61
CAD
CAAAB20956-06
CAAAB20956-14
m-Anisohydrazide 98+%
m-Anisohydrazide
Purity: 98+%
CAS No.: 5785-06-8
Molecular Formula: C8H10N2O2
Formula Weight: 166.18
Melting Point: 90-94°
Beilstein Reference: 2092135
Formula:
C₈H₁₀N₂O₂ Melting Pt: 90…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007601 CAS Number: 5785-06-8 EINECS: 227-310-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...