5-Chloro-2-fluoroaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

B20951-09 B20951-18
CAAAB20951-09EA 106.49 CAD
CAAAB20951-09 CAAAB20951-18
5-Chloro-2-fluoroaniline 97%
5-Chloro-2-fluoroaniline

Purity: 97% CAS No.: 2106-05-0 Molecular Formula: C6H5ClFN Formula Weight: 145.56 Flash Point: 103°(217°F) Refractive Index: 1.5600 Odor: Characteristic Boiling Point: 102-105°/20mm UN No.: 2810 MDL No.: MFCD00069416

Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 102…105 °C/20 mm
Density: 1.30
Flash Pt: 103 °C(217 °F)
Storage Temperature: Ambient
MDL Number: MFCD00069416
CAS Number: 2106-05-0
EINECS: 218-284-1
UN: 2810

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR