3,5-Dichlorosalicylaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20941-22
B20941-06
B20941-14
CAAAAB20941-22EA
511.92
CAD
CAAAAB20941-22
CAAAB20941-06
CAAAB20941-14
3,5-Dichlorosalicylaldehyde 98%
3,5-Dichlorosalicylaldehyde
Purity: 98%
CAS No.: 90-60-8
Molecular Formula: C7H4Cl2O2
Formula Weight: 191.01
Melting Point: 94-97°
Beilstein Reference: 973391
Formula:
C₇H₄Cl₂O₂ Boiling Pt: 723 °C (1013 hPa) Melting Pt: 94…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003320 CAS Number: 90-60-8 EINECS: 202-005-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...