1,2-Dibromo-4-fluorobenzene 98%

Supplier: Thermo Scientific Chemicals

B20935-09 B20935-88
CAAAB20935-09EA 170.64 CAD
CAAAB20935-09 CAAAB20935-88
1,2-Dibromo-4-fluorobenzene 98%
1,2-Dibromo-4-fluorobenzene

Purity: 98% CAS No.: 2369-37-1 Molecular Formula: C6H3Br2F Formula Weight: 253.91 MDL No.: MFCD00070780

Formula: C₆H₃Br₂F
MW: 253.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070780
CAS Number: 2369-37-1
EINECS: 219-131-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR