2,3-Dichlorophenyl isothiocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3-Dichlorophenylisothiocyanate, 1,2-Dichloro-3-isothiocyanatobenzene

B20930-06 B20930-14
CAAAB20930-06EA 64.46 CAD
CAAAB20930-06 CAAAB20930-14
2,3-Dichlorophenyl isothiocyanate 97%
2,3-Dichlorophenyl isothiocyanate

Purity: 97% CAS No.: 6590-97-2 Molecular Formula: C7H3Cl2NS Formula Weight: 204.08 Flash Point: 110°(230°F) Refractive Index: 1.6740 Odor: Irritating Boiling Point: 256-258° Beilstein Reference: 2091585 UN No.: 2922 MDL No.: MFCD00041048

Formula: C₇H₃Cl₂NS
MW: 204.08
Boiling Pt: 256…258 °C
Density: 1.434
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041048
CAS Number: 6590-97-2
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR