1H,1H,2H-Perfluoro-1-octene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20920-06
B20920-14
CAAAB20920-06EA
59.25
CAD
CAAAB20920-06
CAAAB20920-14
1H,1H,2H-Perfluoro-1-octene 99%
1H,1H,2H-Perfluoro-1-octene
Purity: 99%
CAS No.: 25291-17-2
Molecular Formula: CF3(CF2)5CH=CH2
Formula Weight: 346.09
Flash Point: 20°(202°F)
Refractive Index: 1.2960
Boiling Point: 106-107°
Beilstein Reference: 2060994
MDL No.: MFCD00039249
Formula:
C₈H₃F₁₃ MW: 346.09 g/mol Boiling Pt: 106…107 °C Density: 1.580 Flash Pt: 20 °C (202 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039249 CAS Number: 25291-17-2 EINECS: 246-791-8 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...