3-Heptyn-1-ol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20884-06
B20884-14
CAAAB20884-06EA
67.62
CAD
CAAAB20884-06
CAAAB20884-14
3-Heptyn-1-ol 98%
3-Heptyn-1-ol
Purity: 98%
CAS No.: 14916-79-1
Molecular Formula: CH3CH2CH2C≡CCH2CH2OH
Formula Weight: 112.17
Flash Point: 74°(165°F)
Refractive Index: 1.4555
Boiling Point: 80-82°/15mm
Beilstein Reference: 1737178
MDL No.: MFCD00039551
Formula:
C₇H₁₂O Boiling Pt: 80…82 °C/15 mm Density: 0.882 Flash Pt: 74 °C(165 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039551 CAS Number: 14916-79-1 EINECS: 238-985-6 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...