1-Pentynylbenzene 98%
Supplier: Thermo Scientific Chemicals
B20857-06
B20857-14
CAAAB20857-06EA
55.93
CAD
CAAAB20857-06
CAAAB20857-14
1-Pentynylbenzene 98%
1-Pentynylbenzene
Purity: 98%
CAS No.: 4250-81-1
Molecular Formula: C11H12
Formula Weight: 144.22
Flash Point: 82°(179°F)
Refractive Index: 1.5413
Boiling Point: 213-215°
Beilstein Reference: 1853156
MDL No.: MFCD00039959
Formula:
C₆H₅CC(CH₂)₂CH₃ Boiling Pt: 213…215 °C Density: 0.906 |
MDL Number:
MFCD00039959 CAS Number: 4250-81-1 EINECS: 224-211-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...