2-Methylnonane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20849-03
B20849-06
B20849-14
CAAAB20849-03EA
79
CAD
CAAAB20849-03
CAAAB20849-06
CAAAB20849-14
2-Methylnonane 98+%
2-Methylnonane
Purity: 98+%
CAS No.: 871-83-0
Molecular Formula: C10H22
Formula Weight: 142.29
Flash Point: 46°(115°F)
Refractive Index: 1.410
Odor: Petroleum-like
Boiling Point: 166°
Beilstein Reference: 1696990
UN No.: 3295
MDL No.: MFCD00027321
Formula:
C₁₀H₂₂ Boiling Pt: 166 °C Melting Pt: –74.6 °C Density: 0.72 Flash Pt: 46 °C (115 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00027321 CAS Number: 871-83-0 EINECS: 212-814-5 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...