Chloramphenicol 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20841-14
B20841-22
CAAAB20841-14EA
124.69
CAD
CAAAB20841-14
CAAAB20841-22
Chloramphenicol 99+%
Chloramphenicol
Inhibitor of translation on the 50S subunit at the peptidyltransferase step
Purity: 99+%
CAS No.: 56-75-7
Molecular Formula: C11H12Cl2N2O5
Formula Weight: 323.14
Physical Form: Crystalline powder
Appearance: White to pale cream
Melting Point: 149-152°
Beilstein Reference: 2225532
Merck Reference: 14,2077
Optical Rotation: +19° (c=6 in ethanol)
MDL No.: MFCD00078159
Inhibitor of translation on the 50S subunit at the peptidyltransferase step
Formula:
Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂ Boiling Pt: 645 °C (1013 hPa) Melting Pt: 149…152 °C Density: 1.49 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00078159 CAS Number: 56-75-7 EINECS: 200-287-4 Merck Index: 14,2077 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...