2-(Trifluoromethoxy)benzaldehyde 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20840-22
B20840-06
B20840-14
CAAAAB20840-22EA
684.14
CAD
CAAAAB20840-22
CAAAB20840-06
CAAAB20840-14
2-(Trifluoromethoxy)benzaldehyde 96%
2-(Trifluoromethoxy)benzaldehyde
Purity: 96%
CAS No.: 94651-33-9
Molecular Formula: C8H5F3O2
Formula Weight: 190.12
Flash Point: 67°(152°F)
Refractive Index: 1.4550
Boiling Point: 77°/20mm
Beilstein Reference: 6137162
MDL No.: MFCD00042405
Formula:
C₈H₅F₃O₂ Boiling Pt: 77 °C/20 mm Density: 1.330 Flash Pt: 67 °C (152 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042405 CAS Number: 94651-33-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...