p,α-Dimethylbenzyl alcohol 97%
Supplier: Thermo Scientific Chemicals
B20832-03
B20832-06
B20832-14
CAAAB20832-03EA
56.56
CAD
CAAAB20832-03
CAAAB20832-06
CAAAB20832-14
p,α-Dimethylbenzyl alcohol 97%
p,α-Dimethylbenzyl alcohol
Purity: 97%
CAS No.: 536-50-5
Molecular Formula: C9H12O
Formula Weight: 136.20
Flash Point: 107°(225°F)
Refractive Index: 1.5230
Boiling Point: 218-220°
Beilstein Reference: 1857562
Merck Reference: 14,3237
MDL No.: MFCD00016860
Formula:
C₉H₁₂O Boiling Pt: 218…220 °C Melting Pt: 81…82 °C Density: 0.987 Flash Pt: 107 °C(225 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00016860 CAS Number: 536-50-5 EINECS: 208-637-8 Merck Index: 14,3237 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...