5-Fluoroisatin 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Fluoroindoline-2,3-dione, 5-Fluoro-1H-indole-2,3-dione

B20831-14 B20831-03 B20831-06
CAAAAB20831-14EA 377.62 CAD
CAAAAB20831-14 CAAAB20831-03 CAAAB20831-06
5-Fluoroisatin 98%
5-Fluoroisatin

Purity: 98% CAS No.: 443-69-6 Molecular Formula: C8H4FNO2 Formula Weight: 165.12 Melting Point: 224-227° Beilstein Reference: 131241

Formula: C₈H₄FNO₂
MW: 165.12 g/mol
Melting Pt: 224…227 °C
Storage Temperature: Ambient
MDL Number: MFCD00022795
CAS Number: 443-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR