trans-3-Fluorocinnamic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B20818-22 B20818-06 B20818-14
CAAAAB20818-22EA 669.92 CAD
CAAAAB20818-22 CAAAB20818-06 CAAAB20818-14
trans-3-Fluorocinnamic acid 98%
trans-3-Fluorocinnamic acid

Purity: 98% CAS No.: 20595-30-6 Molecular Formula: C9H7FO2 Formula Weight: 166.15 Odor: Odorless Melting Point: 165-168° Beilstein Reference: 1365172 MDL No.: MFCD00004383

Formula: C₉H₇FO₂
Melting Pt: 165…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00004383
CAS Number: 20595-30-6
EINECS: 207-281-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR