2,3-Difluorobenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

B20811-03 B20811-06
CAAAB20811-03EA 50.09 CAD
CAAAB20811-03 CAAAB20811-06
2,3-Difluorobenzonitrile 99%
2,3-Difluorobenzonitrile

Purity: 99% CAS No.: 21524-39-0 Molecular Formula: C7H3F2N Formula Weight: 139.11 Flash Point: 75°(167°F) Refractive Index: 1.4882 Odor: Almond-like Boiling Point: 187-188° Beilstein Reference: 2085621 UN No.: 3276 MDL No.: MFCD00009976

Formula: C₇H₃F₂N
MW: 139.1 g/mol
Boiling Pt: 187…188 °C
Density: 1.254
Flash Pt: 75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009976
CAS Number: 21524-39-0
EINECS: 244-418-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR