3-Nitrobenzhydrazide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20800-06
B20800-14
CAAAB20800-06EA
69.68
CAD
CAAAB20800-06
CAAAB20800-14
3-Nitrobenzhydrazide 98+%
3-Nitrobenzhydrazide
Purity: 98+%
CAS No.: 618-94-0
Molecular Formula: C7H7N3O3
Formula Weight: 181.15
Melting Point: 152-155°
Beilstein Reference: 645512
Formula:
C₇H₇N₃O₃ Melting Pt: 152…155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017059 CAS Number: 618-94-0 EINECS: 210-572-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...