2-Cyclohexylethanol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20782-0B
B20782-18
B20782-30
CAAAB20782-0BEA
685.72
CAD
CAAAB20782-0B
CAAAB20782-18
CAAAB20782-30
2-Cyclohexylethanol 99%
2-Cyclohexylethanol
Purity: 99%
CAS No.: 4442-79-9
Molecular Formula: C8H16O
Formula Weight: 128.22
Flash Point: 86°(186°F)
Refractive Index: 1.4660
Melting Point: -20°
Boiling Point: 206-207°
Beilstein Reference: 1848152
MDL No.: MFCD00001525
Formula:
C₈H₁₆O MW: 128.21 g/mol Boiling Pt: 206…207 °C Melting Pt: –20 °C Density: 0.920 Flash Pt: 86 °C(186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001525 CAS Number: 4442-79-9 EINECS: 224-672-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...