1-Iodopentane 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with copper
B20781-22
B20781-36
CAAAB20781-22EA
100.96
CAD
CAAAB20781-22
CAAAB20781-36
1-Iodopentane 98% stabilized
1-Iodopentane
Purity: 98%
CAS No.: 628-17-1
Molecular Formula: C5H11I
Formula Weight: 198.05
Flash Point: 51°(123°F)
Refractive Index: 1.4950
Odor: Characteristic
Melting Point: -86°
Boiling Point: 154-156°
Beilstein Reference: 1730982
UN No.: 1993
MDL No.: MFCD00001100
Formula:
C₅H₁₁I Boiling Pt: 154…156 °C Melting Pt: –86 °C Density: 1.515 Flash Pt: 51 °C (123 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001100 CAS Number: 628-17-1 EINECS: 211-030-0 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...