2,4,4-Trimethyl-2-pentene 97%

Supplier: Thermo Scientific Chemicals
Danger

B20773-14 B20773-22
CAAAB20773-14EA 195.92 CAD
CAAAB20773-14 CAAAB20773-22
2,4,4-Trimethyl-2-pentene 97%
2,4,4-Trimethyl-2-pentene

Purity: 97% CAS No.: 107-40-4 Molecular Formula: C8H16 Formula Weight: 112.22 Flash Point: -1°(29°F) Refractive Index: 1.4160 Melting Point: -106° Boiling Point: 103-105° UN No.: 3295 MDL No.: MFCD00008902

Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 103…105 °C
Melting Pt: –106 °C
Density: 0.720
Flash Pt: –1 °C (29 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008902
CAS Number: 107-40-4
EINECS: 203-488-5
UN: 2050
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR