Benzalkonium chloride 50% (w/w) in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20760-0F
B20760-AP
CAAAB20760-0FEA
453.46
CAD
CAAAB20760-0F
CAAAB20760-AP
Benzalkonium chloride 50% (w/w) in aqueous solution
Benzalkonium chloride
Cationic surface active agent
CAS No.: 63449-41-2
Flash Point: None
Beilstein Reference: 4062599
Merck Reference: 14,1059
UN No.: 3265
MDL No.: MFCD00145757
Formula:
C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇) Density: 0.99 Storage Temperature: Ambient |
MDL Number:
MFCD00145757 CAS Number: 63449-41-2 EINECS: 264-151-6 UN: 3265 ADR: 8,III Merck Index: 14,1059 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...