(3-Bromophenyl)acetic acid 98+%
Supplier: Thermo Scientific Chemicals
B20758-14
B20758-06
CAAAAB20758-14EA
431.34
CAD
CAAAAB20758-14
CAAAB20758-06
(3-Bromophenyl)acetic acid 98+%
(3-Bromophenyl)acetic acid
Purity: 98+%
CAS No.: 1878-67-7
Molecular Formula: C8H7BrO2
Formula Weight: 215.05
Melting Point: 100-102°
Beilstein Reference: 2045104
Formula:
(C₆H₄Br)CH₂COOH Boiling Pt: 308 °C (1013 hPa) Melting Pt: 100…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004330 CAS Number: 1878-67-7 EINECS: 217-522-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...