α-Bromo-2,5-difluorotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20752-18
B20752-09
CAAAAB20752-18EA
554.58
CAD
CAAAAB20752-18
CAAAB20752-09
α-Bromo-2,5-difluorotoluene 98%
α-Bromo-2,5-difluorotoluene
Purity: 98%
CAS No.: 85117-99-3
Molecular Formula: C7H5BrF2
Formula Weight: 207.02
Refractive Index: 1.5260
Beilstein Reference: 7089248
UN No.: 3265
MDL No.: MFCD00009897
Formula:
C₇H₅BrF₂ Boiling Pt: 186 °C (1013 hPa) Melting Pt: 8 °C Density: 1.609 Storage Temperature: Ambient |
MDL Number:
MFCD00009897 CAS Number: 85117-99-3 EINECS: 285-651-0 UN: 2924 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...