2-Methyl-1-phenyl-1-propanol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: α-Isopropylbenzyl alcohol, 2-Methyl-1-phenylpropan-1-ol

B20729-06 B20729-14
CAAAB20729-06EA 159.58 CAD
CAAAB20729-06 CAAAB20729-14
2-Methyl-1-phenyl-1-propanol 98%
2-Methyl-1-phenyl-1-propanol

Purity: 98% CAS No.: 611-69-8 Molecular Formula: C10H14O Formula Weight: 150.22 Flash Point: 86°(187°F) Refractive Index: 1.5130 Boiling Point: 124-125°/15mm MDL No.: MFCD00065000

Formula: C₆H₅CH(OH)CH(CH₃)₂
Boiling Pt: 124…125 °C/15 mm
Melting Pt: –14 °C
Density: 0.964
Flash Pt: 86 °C(187 °F)
Storage Temperature: Ambient
MDL Number: MFCD00065000
CAS Number: 611-69-8
EINECS: 210-274-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=954485764810b5712627a8e76df6a8dd392a6e2a&vid=954485764810b5712627a8e76df6a8dd392a6e2a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR