Disulfiram 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20721-0B
B20721-0I
B20721-30
CAAAB20721-0BEA
191.18
CAD
CAAAB20721-0B
CAAAB20721-0I
CAAAB20721-30
Disulfiram 97%
Dopamine beta-hydroxylase inhibitor
Purity: 97%
CAS No.: 97-77-8
Molecular Formula: C10H20N2S4
Formula Weight: 296.54
Melting Point: 69-72°
Boiling Point: 117°/17mm
Merck Reference: 14,3364
UN No.: 3077
MDL No.: MFCD00009048
Formula:
C₁₀H₂₀N₂S₄ Boiling Pt: 117 °C/17 mm Melting Pt: 69…72 °C Density: 1.300 Storage Temperature: Ambient |
MDL Number:
MFCD00009048 CAS Number: 97-77-8 EINECS: 202-607-8 UN: 3077 ADR: 9,III Merck Index: 14,3364 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...