Methyl-3-mercaptopropionate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Mercaptopropionic acid methyl ester, Methyl 3-mercaptopropanoate
B20718-14
B20718-22
CAAAB20718-14EA
52.14
CAD
CAAAB20718-14
CAAAB20718-22
Methyl-3-mercaptopropionate 98%
Methyl 3-mercaptopropionate
Purity: 98%
CAS No.: 2935-90-2
Molecular Formula: HSCH2CH2CO2CH3
Formula Weight: 120.17
Flash Point: 60°(140°F)
Refractive Index: 1.4640
Boiling Point: 164°
Beilstein Reference: 1742952
UN No.: 1228
MDL No.: MFCD00004895
Formula:
C₄H₈O₂S MW: 120.17 g/mol Boiling Pt: 164 °C Melting Pt: –60 °C Density: 1.085 Flash Pt: 60 °C (140 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004895 CAS Number: 2935-90-2 EINECS: 220-912-4 UN: 3071 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...