(±)-1-Octen-4-ol 99%

Supplier: Thermo Scientific Chemicals
Warning

B20707-06 B20707-14
CAAAB20707-06EA 118.97 CAD
CAAAB20707-06 CAAAB20707-14
(±)-1-Octen-4-ol 99%
(±)-1-Octen-4-ol

Purity: 99% CAS No.: 40575-42-6 Molecular Formula: C8H16O Formula Weight: 128.22 MDL No.: MFCD00021943

Formula: C₈H₁₆O
Density: 0.83
Storage Temperature: Ambient
MDL Number: MFCD00021943
CAS Number: 40575-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR