2,3,4-Trifluorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B20696-14 B20696-06
CAAAAB20696-14EA 603.56 CAD
CAAAAB20696-14 CAAAB20696-03 CAAAB20696-06
2,3,4-Trifluorobenzoic acid 98%
2,3,4-Trifluorobenzoic acid

Purity: 98% CAS No.: 61079-72-9 Molecular Formula: C7H3F3O2 Formula Weight: 176.10 Melting Point: 140-142° Beilstein Reference: 7476020

Formula: C₇H₃F₃O₂
MW: 176.09 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00061232
CAS Number: 61079-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR