Phenyl salicylate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20686-22
B20686-36
CAAAB20686-22EA
50.56
CAD
CAAAB20686-22
CAAAB20686-36
Phenyl salicylate 99%
Phenyl salicylate
Purity: 99%
CAS No.: 118-55-8
Molecular Formula: C13H10O3
Formula Weight: 214.22
Flash Point: 160°(320°F)
Physical Form: Crystalline powder
Melting Point: 41-45°
Boiling Point: 172-174°/12mm
Beilstein Reference: 393969
Merck Reference: 14,7310
MDL No.: MFCD00002213
Formula:
C₁₃H₁₀O₃ Boiling Pt: 172…174 °C/12 mm Melting Pt: 41…45 °C Density: 1.307 g/cm³ (20 °C) Flash Pt: 160 °C(320 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002213 CAS Number: 118-55-8 EINECS: 204-259-2 UN: 3077 ADR: 9,III Merck Index: 14,7310 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...