Propyl propionate 99%

Supplier: Thermo Scientific Chemicals
Warning

B20680-AK B20680-AU
CAAAB20680-AKEA 76.16 CAD
CAAAB20680-AK CAAAB20680-AU
Propyl propionate 99%
Propyl propionate

Purity: 99% CAS No.: 106-36-5 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 19°(67°F) Refractive Index: 1.3930 Melting Point: -76° Boiling Point: 122-124° Merck Reference: 14,7867 UN No.: 3272 MDL No.: MFCD00009373

Formula: C₆H₁₂O₂
Boiling Pt: 122…124 °C
Melting Pt: –76 °C
Density: 0.881
Flash Pt: 19 °C (67 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009373
CAS Number: 106-36-5
EINECS: 203-389-7
UN: 3272
ADR: 3,III
Merck Index: 14,7867

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR