(2-Methoxyphenoxy)acetic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(2-Methoxyphenoxy)acetic acid

B20672-06 B20672-14
CAAAB20672-06EA 27.81 CAD
CAAAB20672-06 CAAAB20672-14
(2-Methoxyphenoxy)acetic acid 98+%
(2-Methoxyphenoxy)acetic acid

Purity: 98+% CAS No.: 1878-85-9 Molecular Formula: C9H10O4 Formula Weight: 182.18 Melting Point: 118-122° Beilstein Reference: 1874501

Formula: (C₆H₄OCH₃)OCH₂COOH
MW: 182.18 g/mol
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00014352
CAS Number: 1878-85-9
EINECS: 217-526-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR