3-Amino-p-anisic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20669-06
B20669-14
CAAAB20669-06EA
61.15
CAD
CAAAB20669-06
CAAAB20669-14
3-Amino-p-anisic acid 98+%
3-Amino-p-anisic acid
Purity: 98+%
CAS No.: 2840-26-8
Molecular Formula: C8H9NO3
Formula Weight: 167.16
Melting Point: 208-210°
MDL No.: MFCD00002521
Formula:
C₈H₉NO₃ MW: 167.16 Boiling Pt: 335 Melting Pt: 208…210 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002521 CAS Number: 2840-26-8 EINECS: 220-634-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...