α-Bromo-p-toluonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Bromomethyl)benzonitrile, 4-Cyanobenzyl bromide, p-(Bromomethyl)benzonitrile

B20641-06 B20641-14
CAAAB20641-06EA 97.64 CAD
CAAAB20641-06 CAAAB20641-14
α-Bromo-p-toluonitrile 98%
α-Bromo-p-toluonitrile

Purity: 98% CAS No.: 17201-43-3 Molecular Formula: C8H6BrN Formula Weight: 196.05 Melting Point: 113-117° Beilstein Reference: 636717 UN No.: 2923 MDL No.: MFCD00001829

Formula: C₈H₆BrN
MW: 196.05 g/mol
Melting Pt: 115
Storage Temperature: Ambient
MDL Number: MFCD00001829
CAS Number: 17201-43-3
EINECS: 241-246-0
UN: 1759
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR